AC5428452663 Accela ChemBio Inc.

4-Cyclopropylthiazole

  • MDL number MFCD07390348
  • Cas Number : 433217-34-6
  • Molecular Weight : 125.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY023291&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY023291.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart