AC5428452676 Accela ChemBio Inc.

4-(N,N-Dimethylaminomethyl)phenylboronic Acid Pinacol Ester

  • MDL number MFCD04971190
  • Cas Number : 878197-87-6
  • Molecular Weight : 261.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY024492&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY024492.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart