AC5428452681 Accela ChemBio Inc.

Ethyl 2-(1-Naphthyl)acetate

  • MDL number MFCD00021579
  • Cas Number : 2122-70-5
  • Molecular Weight : 214.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025190&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY025190.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart