AC5428452686 Accela ChemBio Inc.

Methyl (S)-2-(Boc-amino)-4-iodobutanoate

  • MDL number MFCD16294352
  • Cas Number : 101650-14-0
  • Molecular Weight : 343.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY025699&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY025699.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart