AC5428452693 Accela ChemBio Inc.

Methyl 3-(1-Bromoethyl)benzoate

  • MDL number MFCD26937555
  • Cas Number : 50603-99-1
  • Molecular Weight : 243.09714

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026627&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY026627.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart