AC5428452697 Accela ChemBio Inc.

2-Chloro-6-hydroxy-4-methoxybenzaldehyde

  • MDL number MFCD23705594
  • Cas Number : 116475-68-4
  • Molecular Weight : 186.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY026709&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY026709.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart