AC5428452706 Accela ChemBio Inc.

N-Isopropyl-1-imidazolecarboxamide

  • MDL number MFCD11099729
  • Cas Number : 75472-66-1
  • Molecular Weight : 153.18174

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027185&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027185.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart