AC5428452711 Accela ChemBio Inc.

N-Boc-4-(bromomethyl)aniline

  • MDL number MFCD11616718
  • Cas Number : 239074-27-2
  • Molecular Weight : 286.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027408&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027408.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart