AC5428452714 Accela ChemBio Inc.

Methyl 3-Cyclopropyl-3-(3-hydroxyphenyl)propanoate

  • MDL number MFCD27935376
  • Cas Number : 1142224-62-1
  • Molecular Weight : 220.26434

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027621&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027621.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart