AC5428452730 Accela ChemBio Inc.

Ethyl 4-Methoxy-1-methyl-2-oxo-1,2,5,6-tetrahydropyridine-3-carboxylate

  • MDL number MFCD27935380
  • Cas Number : 851726-49-3
  • Molecular Weight : 213.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027989&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY027989.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart