AC5428452733 Accela ChemBio Inc.

Methyl 2-Hydroxy-3-(3-hydroxyphenyl)propanoate

  • MDL number MFCD22381395
  • Cas Number : 1508892-33-8
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028024&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY028024.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart