AC5428452737 Accela ChemBio Inc.

1-Ethylpiperidine-2,4-dione

  • MDL number MFCD13179015
  • Cas Number : 99539-36-3
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028108&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY028108.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart