AC5428452810 Accela ChemBio Inc.

3-Hydroxy-1-methylazetidine

  • MDL number MFCD10565802
  • Cas Number : 111043-48-2
  • Molecular Weight : 87.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY029236&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY029236.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart