AC5428452829 Accela ChemBio Inc.

2-Aminopyrrolo[2,1-f][1,2,4]triazin-4(3H)-one

  • MDL number MFCD21364507
  • Cas Number : 159326-75-7
  • Molecular Weight : 150.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY029365&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY029365.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart