AC5428452846 Accela ChemBio Inc.

5-(Hydroxymethyl)pyridine-3-boronic Acid

  • MDL number MFCD09952042
  • Cas Number : 908369-20-0
  • Molecular Weight : 152.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form White to yellow solid
Shipping Temperature +4 ?
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY029925&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY029925.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart