AC5429242340 Accela ChemBio Inc.

2,6-Difluoro-4-(methylsulfonyl)aniline

  • MDL number MFCD24642337
  • Cas Number : 1147557-74-1
  • Molecular Weight : 207.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY030786&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY030786.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart