AC5429242351 Accela ChemBio Inc.

4-Bromo-3-fluoroindole

  • MDL number MFCD24620115
  • Cas Number : 1253926-20-3
  • Molecular Weight : 214.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form Light yellow to green to brown solid or liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY031150&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY031150.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart