AC5429242380 Accela ChemBio Inc.

2-(Boc-aminomethyl)phenol

  • MDL number MFCD11655886
  • Cas Number : 390427-07-3
  • Molecular Weight : 223.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY031532&attname=GoodsCode

Safety Information

hcodes No GHS SYMBOl
MSDS http://pdf.accelachem.com/cn/msds/SY031532.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart