AC5444725905 Accela ChemBio Inc.

N-Phenethylbiguanide

  • MDL number MFCD00242966
  • Cas Number : 114-86-3
  • Molecular Weight : 205.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY031759&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY031759.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart