AC5444725906 Accela ChemBio Inc.

1,3-Dihydroisobenzofuran-5-boronic Acid Pinacol Ester

  • MDL number MFCD22628089
  • Cas Number : 1352037-60-5
  • Molecular Weight : 246.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY031768&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY031768.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart