AC5444725949 Accela ChemBio Inc.

1-Methyl-4,5,6,7-tetrahydro-1H-imidazo[4,5-c]pyridine

  • MDL number MFCD01081938
  • Cas Number : 87673-88-9
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY032476&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY032476.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart