AC5444725952 Accela ChemBio Inc.

Octahydro-2H-pyrano[3,2-c]pyridine

  • MDL number MFCD18073359
  • Cas Number : 71671-81-3
  • Molecular Weight : 141.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY032540&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY032540.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart