AC5444725992 Accela ChemBio Inc.

3-Chloro-7-nitro-1,2-benzisoxazole

  • MDL number MFCD26141868
  • Cas Number : 178747-55-2
  • Molecular Weight : 198.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY032879&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY032879.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart