AC5444726047 Accela ChemBio Inc.

5-Bromo-1-methylpyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD00038053
  • Cas Number : 6327-97-5
  • Molecular Weight : 205.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-bromo-4-hydroxy-1-methyl-2(1H)-pyrimidinone
Inchi Code 1S/C5H5BrN2O2/c1-8-2-3(6)4(9)7-5(8)10/h2H,1H3,(H,7,9,10)
Inchi Key PIVKLKRNXWUHSD-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY033860&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY033860.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart