AC5448900072 Accela ChemBio Inc.

3,4-Dihydroxybenzophenone

  • MDL number MFCD00477203
  • Cas Number : 10425-11-3
  • Molecular Weight : 214.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018338&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P305 + P351 + P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY018338.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart