AC5448900092 Accela ChemBio Inc.

2-Amino-5-(benzyloxy)-4-methoxybenzonitrile

  • MDL number MFCD16619280
  • Cas Number : 192869-57-1
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY028896&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY028896.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart