AC5448900138 Accela ChemBio Inc.

2-Chloro-5-(methoxymethyl)pyrimidine

  • MDL number MFCD28385449
  • Cas Number : 1416366-34-1
  • Molecular Weight : 158.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY034069&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY034069.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart