AC5448900192 Accela ChemBio Inc.

(1R,3S)-1-(Boc-amino)-3-(Cbz-amino)cyclopentane

  • MDL number MFCD28398154
  • Cas Number : 774212-79-2
  • Molecular Weight : 334.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY035237&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY035237.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart