AC5448900223 Accela ChemBio Inc.

2-Isobutylaniline

  • MDL number MFCD00130034
  • Cas Number : 71182-59-7
  • Molecular Weight : 149.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY036357&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY036357.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart