AC5457496431 Accela ChemBio Inc.

1,2-Dimethoxy-4,5-dinitrobenzene

  • MDL number MFCD00082695
  • Cas Number : 3395-03-7
  • Molecular Weight : 228.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. RT
Physical Form Light yellow to dark yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY032601&attname=GoodsCode

Safety Information

signalword Danger
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501a
hcodes GHS06
MSDS http://pdf.accelachem.com/cn/msds/SY032601.pdf
hstatements H302-H312-H331-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart