AC5457496494 Accela ChemBio Inc.

4-Chloroquinazoline-8-carbonitrile

  • MDL number MFCD15527394
  • Cas Number : 1231761-54-8
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY036304&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY036304.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart