AC5465191319 Accela ChemBio Inc.

1-Benzyl-6-oxopiperidine-3-carboxylic Acid

  • MDL number MFCD17041934
  • Cas Number : 32749-55-6
  • Molecular Weight : 233.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY037266&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY037266.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart