AC5465191330 Accela ChemBio Inc.

6-Hydrazinopyridazine-3-carboxamide

  • MDL number MFCD16698821
  • Cas Number : 3614-47-9
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY037384&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY037384.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart