AC5465191338 Accela ChemBio Inc.

4-Chloro-7-hydroxyquinazoline

  • MDL number MFCD08276177
  • Cas Number : 849345-42-2
  • Molecular Weight : 180.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY037514&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY037514.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart