AC5465191377 Accela ChemBio Inc.

4-(Hydroxymethyl)oxazolidin-2-one

  • MDL number MFCD11045779
  • Cas Number : 15546-08-4
  • Molecular Weight : 117.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038213&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY038213.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart