AC5497683160 Accela ChemBio Inc.

(1R,2S,3S,4S,6R)-rel-5-Oxotricyclo[2.2.1.02,6]heptane-3-carboxylic Acid

  • MDL number MFCD00238753
  • Cas Number : 52730-40-2
  • Molecular Weight : 152.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY027170&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY027170.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart