AC5497683248 Accela ChemBio Inc.

4-(Aminomethyl)-5-methylisoxazole

  • MDL number MFCD16042669
  • Cas Number : 139458-29-0
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY035076&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY035076.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart