AC5497683316 Accela ChemBio Inc.

Methyl 12-Bromododecanoate

  • MDL number MFCD01838179
  • Cas Number : 26825-95-6
  • Molecular Weight : 293.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038725&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY038725.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart