AC5497683328 Accela ChemBio Inc.

2-(4-Chlorophenyl)-5'-fluoro-2'-hydroxyacetophenone

  • MDL number MFCD03094383
  • Cas Number : 527751-48-0
  • Molecular Weight : 264.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY038911&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY038911.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart