AC5497683391 Accela ChemBio Inc.

N-Boc-N'-methylthiourea

  • MDL number MFCD16036320
  • Cas Number : 887913-52-2
  • Molecular Weight : 190.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY040103&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY040103.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart