AC5497683404 Accela ChemBio Inc.

2-Bromo-2-fluoroacetic Acid

  • MDL number MFCD07784239
  • Cas Number : 359-25-1
  • Molecular Weight : 156.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY040308&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY040308.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart