AC5497683408 Accela ChemBio Inc.

Ethyl 7-Bromo-3-oxo-3,4-dihydroquinoxaline-2-carboxylate

  • MDL number MFCD08234668
  • Cas Number : 223711-74-8
  • Molecular Weight : 297.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY040386&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY040386.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart