AC5497683414 Accela ChemBio Inc.

4,6-Dichloroindole-2-carboxylic Acid

  • MDL number MFCD00209870
  • Cas Number : 101861-63-6
  • Molecular Weight : 230.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY040441&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY040441.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart