AC5638472384 Accela ChemBio Inc.

Potassium Cyanate

  • MDL number MFCD00011411
  • Cas Number : 590-28-3
  • Molecular Weight : 81.115

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to off white solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY006099&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY006099.pdf
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart