AC5638472688 Accela ChemBio Inc.

Ethenesulfonyl Chloride

  • MDL number MFCD19200056
  • Cas Number : 6608-47-5
  • Molecular Weight : 126.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form Colorless to yellow liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY030716&attname=GoodsCode

Safety Information

signalword Danger
pcodes P280-P305+P351+P338-P337+P313-P403+P235-
MSDS http://pdf.accelachem.com/cn/msds/SY030716.pdf
hstatements H302-H315-H318-H335-H314
Availability details for
Quantity
  • Availability
  • Add to Cart