AC5638472754 Accela ChemBio Inc.

2,6-Diisopropylbenzaldehyde

  • MDL number MFCD24675884
  • Cas Number : 179554-06-4
  • Molecular Weight : 190.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY036091&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY036091.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart