AC5638472811 Accela ChemBio Inc.

3-(2,5-Dimethyl-1-pyrrolyl)phenol

  • MDL number MFCD01314064
  • Cas Number : 97608-33-8
  • Molecular Weight : 187.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY041430&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY041430.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart