AC5638472884 Accela ChemBio Inc.

Methyl 2-Bromo-1-methylimidazole-5-carboxylate

  • MDL number MFCD02179563
  • Cas Number : 120781-02-4
  • Molecular Weight : 219.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY045562&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280-P302+P352-P301+P312-P304+P340-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY045562.pdf
hstatements H312-H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart