AC5638472934 Accela ChemBio Inc.

N-Ethyl-4-methoxybenzylamine Hydrochloride

  • MDL number MFCD07108568
  • Cas Number : 90389-68-7
  • Molecular Weight : 201.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY046062&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY046062.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart