AC5638472946 Accela ChemBio Inc.

5-[2-(1-Boc-4-piperidyl)-2-oxoethyl]-5H-imidazo[5,1-a]isoindole

  • MDL number MFCD29919355
  • Cas Number : 1402838-14-5
  • Molecular Weight : 381.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY046109&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY046109.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart